4-Chloro-2'-methoxybenzophenone ≥97%
Supplier: Matrix Scientific
086517-1G
086517-2G
086517-5G
102849-016EA
489.23
USD
102849-016
102849-018
102849-020
4-Chloro-2'-methoxybenzophenone ≥97%
4-Chloro-2'-methoxybenzophenone
Hazard: Irritant
Formula:
C₁₄H₁₁ClO₂ Melting Pt: 83…85 °C |
MDL Number:
MFCD01311535 CAS Number: 78589-10-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...