4-Chloro-3',5'-difluorobenzophenone ≥97%

Supplier: Matrix Scientific

086516-1G 086516-2G 086516-5G
102849-010EA 387.69 USD
102849-010 102849-012 102849-014
4-Chloro-3',5'-difluorobenzophenone ≥97%
4-Chloro-3',5'-difluorobenzophenone

Hazard: Irritant

Formula: C₁₃H₇ClF₂O
MDL Number: MFCD01319935
CAS Number: 746651-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR