2-Bromo-4'-chloro-5-methoxybenzophenone ≥97%
Supplier: Matrix Scientific
086515-1G
086515-2G
086515-5G
102849-004EA
490.77
USD
102849-004
102849-006
102849-008
2-Bromo-4'-chloro-5-methoxybenzophenone ≥97%
2-Bromo-4'-chloro-5-methoxybenzophenone
Hazard: Irritant
Formula:
C₁₄H₁₀BrClO₂ Melting Pt: 81…83 °C |
MDL Number:
MFCD00672041 CAS Number: 746651-89-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...