2-Bromo-4'-fluoro-5-methoxybenzophenone ≥97%

Supplier: Matrix Scientific

086514-1G 086514-2G 086514-5G
102848-998EA 490.77 USD
102848-998 102849-000 102849-002
2-Bromo-4'-fluoro-5-methoxybenzophenone ≥97%
2-Bromo-4'-fluoro-5-methoxybenzophenone

Hazard: Irritant

Formula: C₁₄H₁₀BrFO₂
MW: 309.14 g/mol
Melting Pt: 69…71 °C
MDL Number: MFCD00672040
CAS Number: 746651-88-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR