2-Bromo-5-methoxybenzophenone ≥97%

Supplier: Matrix Scientific

Synonyms: (2-Bromo-5-methoxyphenyl)(phenyl)methanone

086513-1G 086513-2G 086513-5G
102848-992EA 418.46 USD
102848-992 102848-994 102848-996
2-Bromo-5-methoxybenzophenone ≥97%
2-Bromo-5-methoxybenzophenone

Hazard: Irritant

Formula: C₁₄H₁₁BrO₂
MW: 291.15 g/mol
Melting Pt: 45…46 °C
MDL Number: MFCD00672036
CAS Number: 60080-98-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR