2-Carboethoxybenzophenone ≥97%

Supplier: Matrix Scientific

086511-1G 086511-2G 086511-5G
102848-980EA 490.77 USD
102848-980 102848-982 102848-984
2-Carboethoxybenzophenone ≥97%
2-Carboethoxybenzophenone

Hazard: Irritant

Formula: C₁₆H₁₄O₃
MDL Number: MFCD00027809
CAS Number: 604-61-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR