4-Bromo-2'-carboethoxybenzophenone ≥97%

Supplier: Matrix Scientific

086510-1G 086510-2G 086510-5G
102848-974EA 490.77 USD
102848-974 102848-976 102848-978
4-Bromo-2'-carboethoxybenzophenone ≥97%
4-Bromo-2'-carboethoxybenzophenone

Hazard: Irritant

Formula: C₁₆H₁₃BrO₃
MDL Number: MFCD00672028
CAS Number: 51476-11-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR