3-Bromo-2'-carboethoxybenzophenone ≥97%

Supplier: Matrix Scientific

086509-1G 086509-2G 086509-5G
102848-968EA 490.77 USD
102848-968 102848-970 102848-972
3-Bromo-2'-carboethoxybenzophenone ≥97%
3-Bromo-2'-carboethoxybenzophenone

Hazard: Irritant

Formula: C₁₆H₁₃BrO₃
MDL Number: MFCD00672027
CAS Number: 746651-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR