4-Bromo-4'-carboethoxybenzophenone ≥97%

Supplier: Matrix Scientific

086507-1G 086507-2G 086507-5G
102848-956EA 490.77 USD
102848-956 102848-958 102848-960
4-Bromo-4'-carboethoxybenzophenone ≥97%
4-Bromo-4'-carboethoxybenzophenone

Hazard: Irritant

Formula: C₁₆H₁₃BrO₃
MDL Number: MFCD00672024
CAS Number: 746651-80-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR