2-Bromo-4'-carboethoxybenzophenone ≥97%

Supplier: Matrix Scientific

086506-1G 086506-2G 086506-5G
102848-950EA 410.62 USD
102848-950 102848-952 102848-954
2-Bromo-4'-carboethoxybenzophenone ≥97%
2-Bromo-4'-carboethoxybenzophenone

Hazard: Irritant

Formula: C₁₆H₁₃BrO₃
MDL Number: MFCD00672022
CAS Number: 746651-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR