2,5-Dibromo-3-dodecylthiophene ≥97%

Supplier: Matrix Scientific

086503-25G 086503-50G
102848-934EA 5233.85 USD
102848-934 102848-936
2,5-Dibromo-3-dodecylthiophene ≥97%
2,5-Dibromo-3-dodecylthiophene

Hazard: Irritant

Formula: C₁₆H₂₆Br₂S
MW: 410.25 g/mol
MDL Number: MFCD00274315
CAS Number: 148256-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR