2,5-Dibromo-3-hexylthiophene ≥97%

Supplier: Matrix Scientific

086500-25G 086500-50G
102848-922EA 5233.85 USD
102848-922 102848-924
2,5-Dibromo-3-hexylthiophene ≥97%
2,5-Dibromo-3-hexylthiophene

Hazard: Irritant/Refrigerate

Formula: C₁₀H₁₄Br₂S
MW: 326.0912 g/mol
Density: 1.521
MDL Number: MFCD00274313
CAS Number: 116971-11-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR