2,5-Dibromo-3-butylthiophene ≥97%

Supplier: Matrix Scientific

086499-25G 086499-50G
102848-918EA 5233.85 USD
102848-918 102848-920
2,5-Dibromo-3-butylthiophene ≥97%
2,5-Dibromo-3-butylthiophene

Hazard: Irritant/Refrigerate

Formula: C₈H₁₀Br₂S
MW: 298.04 g/mol
Boiling Pt: 270-281°C
Density: 1.67
Flash Pt: 110°C
MDL Number: MFCD00672012
CAS Number: 116971-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR