3-Dodecylthiophene ≥97%
Supplier: Matrix Scientific
086498-25G
086498-50G
102848-914EA
1021.54
USD
102848-914
102848-916
3-Dodecylthiophene ≥97%
3-Dodecylthiophene
Hazard: Irritant
Formula:
C₁₆H₂₈S MW: 252.46 g/mol Boiling Pt: 290°C Density: 0.902 |
MDL Number:
MFCD00130121 CAS Number: 104934-52-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...