3-Hexylthiophene ≥97%
Supplier: Matrix Scientific
086495-25G
086495-50G
102848-902EA
760
USD
102848-902
102848-904
3-Hexylthiophene ≥97%
3-Hexylthiophene
Hazard: Flammable/Refrigerate/Stored under Argon
Formula:
C₁₀H₁₆S MW: 168.3 g/mol Boiling Pt: 65 °C (0.45 mmHg) Density: 0.936 Flash Pt: 37 °C |
MDL Number:
MFCD00143181 CAS Number: 1693-86-3 UN: 1993 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...