trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Methyl (1R,4R)-4-(hydroxymethyl)cyclohexane-1-carboxylate, Methyl trans-4-hydroxymethylcyclohexanecarboxylate, (1R,4R)-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate

086471-1G 086471-5G
102848-886EA 152.31 USD
102848-886 102848-890
trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate ≥97%
trans-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate

Hazard: Irritant

Formula: C₉H₁₆O₃
MW: 172.22 g/mol
MDL Number: MFCD03840884
CAS Number: 110928-44-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR