cis-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate ≥97%
Supplier: Matrix Scientific
Synonyms:
Methyl cis-4-hydroxymethylcyclohexane-1-carboxylate
086470-1G
086470-2G
086470-5G
102848-880EA
915.38
USD
102848-880
102848-882
102848-884
cis-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate ≥97%
cis-Methyl 4-(hydroxymethyl)cyclohexanecarboxylate
Hazard: Irritant/Refrigerate
Formula:
C₉H₁₆O₃ MW: 172.23 g/mol |
MDL Number:
MFCD06799220 CAS Number: 13380-85-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...