trans-2-[2-(2-Methoxyphenyl)-2-oxoethyl]cyclohexane-1-carboxylic acid ≥95%
Supplier: Matrix Scientific
086467-1G
086467-5G
086467-10G
102848-862EA
1992.31
USD
102848-862
102848-864
102848-866
trans-2-[2-(2-Methoxyphenyl)-2-oxoethyl]cyclohexane-1-carboxylic acid ≥95%
trans-2-[2-(2-Methoxyphenyl)-2-oxoethyl]cyclohexane-1-carboxylic acid
Hazard: Irritant
Formula:
C₁₆H₂₀O₄ Melting Pt: 109…111 °C |
MDL Number:
MFCD01311383 CAS Number: 735274-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...