cis-3-(4-Fluorobenzoyl)cyclohexane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086461-1G 086461-10G
102848-826EA 795.38 USD
102848-826 102848-830
cis-3-(4-Fluorobenzoyl)cyclohexane-1-carboxylic acid ≥95%
cis-3-(4-Fluorobenzoyl)cyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₅FO₃
Melting Pt: 124…127 °C
MDL Number: MFCD01311280
CAS Number: 735269-81-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR