cis-3-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086459-1G 086459-10G
102848-814EA 795.38 USD
102848-814 102848-818
cis-3-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid ≥95%
cis-3-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₈O₄
MDL Number: MFCD01311281
CAS Number: 735269-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR