trans-2-Benzoylcyclohexane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086455-1G 086455-10G
102848-790EA 795.38 USD
102848-790 102848-794
trans-2-Benzoylcyclohexane-1-carboxylic acid ≥95%
trans-2-Benzoylcyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₆O₃
Melting Pt: 149…151 °C
MDL Number: MFCD00800520
CAS Number: 3586-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR