cis-2-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086453-1G 086453-10G
102848-778EA 998.46 USD
102848-778 102848-782
cis-2-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid ≥95%
cis-2-(2-Methoxybenzoyl)cyclohexane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₅H₁₈O₄
Melting Pt: 141…149 °C
MDL Number: MFCD01311249
CAS Number: 733742-61-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR