trans-2-Carbomethoxycyclohexane-1-carboxyic acid ≥97%

Supplier: Matrix Scientific

086448-1G 086448-10G
102848-748EA 496.92 USD
102848-748 102848-752
trans-2-Carbomethoxycyclohexane-1-carboxyic acid ≥97%
trans-2-Carbomethoxycyclohexane-1-carboxyic acid

Hazard: Irritant

Formula: C₉H₁₄O₄
MW: 186.21 g/mol
Melting Pt: 92…95 °C
MDL Number: MFCD01311243
CAS Number: 2484-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR