Methyl trans-2-hydroxymethylcyclopentane-1-carboxylate ≥95%
Supplier: Matrix Scientific
086444-1G
086444-5G
102848-724EA
1269.23
USD
102848-724
102848-728
Methyl trans-2-hydroxymethylcyclopentane-1-carboxylate ≥95%
Methyl trans-2-hydroxymethylcyclopentane-1-carboxylate
Hazard: Store in Freezer
Formula:
C₈H₁₄O₃ |
MDL Number:
MFCD06799097 CAS Number: 502650-66-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...