trans-2-[2-(2-Methoxyphenyl)-2-oxoethyl]cyclopentane-1-carboxylic acid ≥95%
Supplier: Matrix Scientific
086440-1G
086440-5G
102848-700EA
3256.92
USD
102848-700
102848-704
trans-2-[2-(2-Methoxyphenyl)-2-oxoethyl]cyclopentane-1-carboxylic acid ≥95%
trans-2-[2-(2-Methoxyphenyl)-2-oxoethyl]cyclopentane-1-carboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₈O₄ MW: 262.31 g/mol Melting Pt: 83…86 °C |
MDL Number:
MFCD01311222 CAS Number: 733740-55-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...