cis-2-(3-Methylbenzoyl)cyclopentane-1-carboxylic acid ≥95%

Supplier: Matrix Scientific

086436-1G 086436-5G
102848-676EA 1203.08 USD
102848-676 102848-680
cis-2-(3-Methylbenzoyl)cyclopentane-1-carboxylic acid ≥95%
cis-2-(3-Methylbenzoyl)cyclopentane-1-carboxylic acid

Hazard: Irritant

Formula: C₁₄H₁₆O₃
MDL Number: MFCD01311193
CAS Number: 732253-41-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR