cis-1,2-Cyclopentanedicarboxylic acid ≥97%

Supplier: Matrix Scientific

Synonyms: cis-Cyclopentane-1,2-dicarboxylic acid

086427-1G 086427-10G
102848-622EA 490.77 USD
102848-622 102848-626
cis-1,2-Cyclopentanedicarboxylic acid ≥97%
cis-1,2-Cyclopentanedicarboxylic acid

Hazard: Irritant

Formula: C₇H₁₀O₄
MW: 158.16 g/mol
MDL Number: MFCD01311172
CAS Number: 1461-96-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR