Ethyl thiophenene-3-glyoxylate ≥97%

Supplier: Matrix Scientific

086422-1G 086422-5G
102848-592EA 530.77 USD
102848-592 102848-596
Ethyl thiophenene-3-glyoxylate ≥97%
Ethyl thiophenene-3-glyoxylate

Hazard: Irritant

Formula: C₈H₈O₃S
MDL Number: MFCD09801444
CAS Number: 53091-09-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR