Ethyl-3-methylthiophene-2-glyoxylate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(3-methylthiophen-2-yl)-2-oxoacetate

086420-1G 086420-5G
102848-580EA 530.77 USD
102848-580 102848-584
Ethyl-3-methylthiophene-2-glyoxylate ≥97%
Ethyl-3-methylthiophene-2-glyoxylate

Hazard: Irritant

Formula: C₉H₁₀O₃S
MDL Number: MFCD07782945
CAS Number: 32977-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR