Ethyl-9-phenanthroylformate ≥97%

Supplier: Matrix Scientific

086412-1G 086412-5G
102848-532EA 530.77 USD
102848-532 102848-536
Ethyl-9-phenanthroylformate ≥97%
Ethyl-9-phenanthroylformate

Hazard: Irritant

Formula: C₁₈H₁₄O₃
MDL Number: MFCD09801437
CAS Number: 139746-29-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR