Ethyl-4-methyl-1-naphthoylformate ≥97%
Supplier: Matrix Scientific
086410-1G
086410-5G
102848-520EA
530.77
USD
102848-520
102848-524
Ethyl-4-methyl-1-naphthoylformate ≥97%
Ethyl-4-methyl-1-naphthoylformate
Hazard: Irritant
Formula:
C₁₅H₁₄O₃ MW: 242.28 g/mol |
MDL Number:
MFCD09801435 CAS Number: 101093-81-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...