Ethyl-4-fluoro-2-methylbenzoylformate ≥97%

Supplier: Matrix Scientific

086402-1G 086402-5G
102848-472EA 530.77 USD
102848-472 102848-476
Ethyl-4-fluoro-2-methylbenzoylformate ≥97%
Ethyl-4-fluoro-2-methylbenzoylformate

Hazard: Irritant

Formula: C₁₁H₁₁FO₃
MDL Number: MFCD09801426
CAS Number: 334477-07-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR