Ethyl-5-fluoro-2-methoxybenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 5-fluoro-2-methoxybenzoylformate, Ethyl 2-(5-fluoro-2-methoxyphenyl)-2-oxoacetate

086398-1G 086398-5G
102848-448EA 530.77 USD
102848-448 102848-452
Ethyl-5-fluoro-2-methoxybenzoylformate ≥97%
Ethyl-5-fluoro-2-methoxybenzoylformate

Hazard: Irritant/Refrigerate

Formula: C₁₁H₁₁FO₄
MW: 226.21 g/mol
MDL Number: MFCD09801422
CAS Number: 873548-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR