Ethyl-3,4-dimethoxybenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(3,4-dimethoxyphenyl)-2-oxoacetate

086381-1G 086381-5G
102848-346EA 833.85 USD
102848-346 102848-350
Ethyl-3,4-dimethoxybenzoylformate ≥97%
Ethyl 3,4-dimethoxybenzoylformate

Hazard: Irritant

Formula: C₁₂H₁₄O₅
Melting Pt: 39…41 °C
CAS Number: 40233-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR