Ethyl-2,4-dimethoxybenzoylformate ≥97%

Supplier: Matrix Scientific

086379-1G 086379-5G
102848-334EA 981.54 USD
102848-334 102848-338
Ethyl-2,4-dimethoxybenzoylformate ≥97%
Ethyl-2,4-dimethoxybenzoylformate

Hazard: Refrigerate

Formula: C₁₂H₁₄O₅
MDL Number: MFCD09801404
CAS Number: 6592-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR