Ethyl-3,5-dichlorobenzoylformate ≥97%
Supplier: Matrix Scientific
086377-1G
086377-5G
102848-322EA
530.77
USD
102848-322
102848-326
Ethyl-3,5-dichlorobenzoylformate ≥97%
Ethyl 3,5-dichlorobenzoylformate
Hazard: Irritant
Formula:
C₁₀H₈Cl₂O₃ |
MDL Number:
MFCD06201700 CAS Number: 845790-52-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...