Ethyl-2,4-dichlorobenzoylformate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2,4-dichlorobenzoylformate
086375-1G
086375-5G
102848-310EA
530.77
USD
102848-310
102848-314
Ethyl-2,4-dichlorobenzoylformate ≥97%
Ethyl-2,4-dichlorobenzoylformate
Hazard: Irritant
Formula:
C₁₀H₈Cl₂O₃ MW: 247.08 g/mol |
MDL Number:
MFCD03094603 CAS Number: 34966-51-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...