Ethyl-4-chloro-2-fluorobenzoylformate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(4-chloro-2-fluorophenyl)-2-oxoacetate
086367-1G
086367-5G
102848-262EA
530.77
USD
102848-262
102848-266
Ethyl-4-chloro-2-fluorobenzoylformate ≥97%
Ethyl-4-chloro-2-fluorobenzoylformate
Hazard: Irritant/Refrigerate
Formula:
C₁₀H₈ClFO₃ MW: 230.62 g/mol |
MDL Number:
MFCD09801394 CAS Number: 228998-72-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...