Ethyl-3-chloro-4-fluorobenzoylformate ≥97%
Supplier: Matrix Scientific
086365-1G
086365-5G
102848-250EA
530.77
USD
102848-250
102848-254
Ethyl-3-chloro-4-fluorobenzoylformate ≥97%
Ethyl 3-chloro-4-fluorobenzoylformate
Hazard: Irritant
Formula:
C₁₀H₈ClFO₃ |
CAS Number:
845790-53-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...