Ethyl-4-butylbenzoylformate ≥97%

Supplier: Matrix Scientific

086361-1G 086361-5G
102848-226EA 387.69 USD
102848-226 102848-230
Ethyl-4-butylbenzoylformate ≥97%
Ethyl 4-butylbenzoylformate

Hazard: Irritant

Formula: C₁₄H₁₈O₃
MW: 234.291 g/mol
CAS Number: 80120-35-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR