Ethyl-2-phenylbenzoylformate ≥97%

Supplier: Matrix Scientific

086357-1G 086357-5G
102848-202EA 530.77 USD
102848-202 102848-206
Ethyl-2-phenylbenzoylformate ≥97%
Ethyl-2-phenylbenzoylformate

Hazard: Irritant/Refrigerate

Formula: C₁₆H₁₄O₃
MW: 254.29 g/mol
MDL Number: MFCD09801386
CAS Number: 947701-96-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR