Ethyl-3-bromobenzoyl formate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(3-bromophenyl)-2-oxoacetate
086353-1G
086353-5G
102848-178EA
907.69
USD
102848-178
102848-182
Ethyl-3-bromobenzoyl formate ≥97%
Ethyl-3-bromobenzoyl formate
Hazard: Irritant
Formula:
C₁₀H₉BrO₃ MW: 257.09 g/mol |
MDL Number:
MFCD07698676 CAS Number: 62123-80-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...