Ethyl-1-naphthoylformate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(naphthalen-1-yl)-2-oxoacetate, Ethyl (naphth-1-yl)(oxo)acetate
086351-1G
086351-5G
102848-166EA
592.31
USD
102848-166
102848-170
Ethyl-1-naphthoylformate ≥97%
Ethyl-1-naphthoylformate
Hazard: Irritant/Refrigerate
Formula:
C₁₄H₁₂O₃ MW: 228.25 g/mol |
MDL Number:
MFCD01319637 CAS Number: 33656-65-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...