Ethyl-3-methylbenzoylformate ≥96%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-oxo-2-(m-tolyl)acetate
086349-1G
086349-5G
102848-154EA
387.69
USD
102848-154
102848-158
Ethyl-3-methylbenzoylformate ≥96%
Ethyl 3-methylbenzoylformate
Hazard: Irritant/Refrigerate
Formula:
C₁₁H₁₂O₃ |
MDL Number:
MFCD01311168 CAS Number: 66644-68-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...