Diethyl-2-oxooctane-1,8-dicarboxylate ≥95%

Supplier: Matrix Scientific

086345-1G 086345-5G
102848-130EA 981.54 USD
102848-130 102848-134
Diethyl-2-oxooctane-1,8-dicarboxylate ≥95%
Diethyl-2-oxooctane-1,8-dicarboxylate

Hazard: Irritant/Refrigerate

Formula: C₁₂H₂₀O₅
MDL Number: MFCD01319631
CAS Number: 856065-92-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR