Ethyl-4-cyano-2-oxobutyrate ≥97%

Supplier: Matrix Scientific

086340-1G 086340-5G
102848-106EA 907.69 USD
102848-106 102848-110
Ethyl-4-cyano-2-oxobutyrate ≥97%
Ethyl-4-cyano-2-oxobutyrate

Hazard: Irritant/Refrigerate

Formula: C₇H₉NO₃
MDL Number: MFCD01319626
CAS Number: 152831-97-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR