Ethyl-3-fluoro-4-methylbenzoylformate ≥97%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-(3-fluoro-4-methylphenyl)-2-oxoacetate
086338-1G
086338-5G
102848-094EA
387.69
USD
102848-094
102848-098
Ethyl-3-fluoro-4-methylbenzoylformate ≥97%
Ethyl 3-fluoro-4-methylbenzoylformate
Hazard: Irritant/Refrigerate
Formula:
C₁₁H₁₁FO₃ |
MDL Number:
MFCD01319622 CAS Number: 732251-65-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...