Ethyl-4-fluorobenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(4-fluorophenyl)-2-oxoacetate

086336-1G 086336-5G
102848-082EA 387.69 USD
102848-082 102848-086
Ethyl-4-fluorobenzoylformate ≥97%
Ethyl 4-fluorobenzoylformate

Hazard: Irritant/Refrigerate

Formula: C₁₀H₉FO₃
MW: 196.1751 g/mol
MDL Number: MFCD01319617
CAS Number: 1813-94-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR