Ethyl-3-methoxybenzoylformate ≥97%

Supplier: Matrix Scientific

Synonyms: Ethyl 2-(3-methoxyphenyl)-2-oxoacetate

086327-1G 086327-5G
102848-028EA 606.15 USD
102848-028 102848-032
Ethyl-3-methoxybenzoylformate ≥97%
Ethyl 3-methoxybenzoylformate

Hazard: Irritant/Refrigerate

Formula: C₁₀H₉ClO₃
CAS Number: 86358-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR