4-(3-Pyridyl)aniline ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Pyridin-3-ylaniline
086325-1G
086325-5G
102848-016EA
592.31
USD
102848-016
102848-020
4-(3-Pyridyl)aniline ≥97%
4-(3-Pyridyl)aniline
Hazard: Irritant
Formula:
C₁₁H₁₀N₂ MW: 170.22 g/mol Melting Pt: 112…115 °C |
MDL Number:
MFCD00956763 CAS Number: 82261-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...